Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939825_s1
          See other 4932434E15RIK GE Assays ›
          Gene Symbol:
          4932434E15Rik
          Gene Name:
          RIKEN cDNA 4932434E15 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939825_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4932434E15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016545.1 - 1 2752 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream prostate transmembrane protein,
          androgen induced 1
          Pmepa1 2210418I02Rik,
          AW455466,
          Erg1.2,
          N4wbp4,
          Stag1,
          Tmepai
          -19.0kb Anti-sense Chr. 2 - 173224458 - 173276533 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700021F07 gene 1700021F07Rik 223.6kb Sense Chr. 2 - 173522592 - 173528502 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394832 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132378 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132378 - -
          Whole Mouse Genome G4122A A_51_P123965 AK016545.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108418 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01231896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01603569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02215806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01343284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01662159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01682796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01343280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01682794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01343278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01343288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01684399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01684396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868971 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431794_PM_at - -
          Mouse Genome 430 2.0 Array 1431794_PM_at - -
          Mouse Genome 430 2.0 Array 1431794_at - -
          Mouse Expression Set 430 1431794_at - -
          Mouse Expression Set 430 1431794_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431794_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431794_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555108 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.129.208:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix