Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939897_g1
          See other KCNMB4OS2 GE Assays ›
          Gene Symbol:
          Kcnmb4os2
          Gene Name:
          potassium large conductance calcium-activated channel, subfamily M, beta member 4, opposite strand 2
          Gene Aliases:
          4933400F03Rik
          Chromosome Location:
          Chr.10: 116473163 - 116474987 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939897_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnmb4os2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016591.1 - 2 - 3 1369 98
          AW125814.1 - 1 - 2 98
          BF472784.1 - 1 - 2 285 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE945710.1 - 1 - 2 Chr. 10 - 116474421 - 116474989 [-] on Build GRCm38 98
          GenBankmRNA BE980425.1 - 1 - 2 Chr. 10 - 116474369 - 116474989 [-] on Build GRCm38 98
          GenBankmRNA BE979773.1 - 1 - 2 Chr. 10 - 116474422 - 116474989 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream potassium large conductance calcium-activated channel,
          subfamily M,
          beta member 4
          Kcnmb4 1700058G18Rik,
          2900045G12Rik
          -603.0 Anti-sense Chr. 10 - 116417868 - 116473882 on NCBI Build GRCm38
          Downstream CCR4-NOT transcription complex,
          subunit 2
          Cnot2 2600016M12Rik,
          2810470K03Rik,
          AA537049,
          AA959607,
          AW557563,
          C79650
          10.2kb Anti-sense Chr. 10 - 116485161 - 116581535 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2306336 AK016591.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657450 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138469 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2306336 AK016591.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138469 - -
          Whole Mouse Genome G4122A A_51_P488819 AK016591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488819 AK165712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488819 AW125814.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125738 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454079_PM_at - -
          Mouse Genome 430 2.0 Array 1454079_at - -
          Mouse Genome 430 2.0 Array 1454079_PM_at - -
          Mouse Expression Set 430 1454079_at - -
          Mouse Expression Set 430 1454079_PM_at - -
          Murine Genome U74v2 Set 138425_at - -
          Murine Genome U74v2 Set 138425_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454079_at - -
          Affymetrix HT Mouse Genome 430B Array 1454079_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0