Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939980_s1
          See other 4933407A17RIK GE Assays ›
          Gene Symbol:
          4933407A17Rik
          Gene Name:
          RIKEN cDNA 4933407A17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939980_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933407A17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016710.1 - 1 749 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphodiesterase 12 Pde12 2'-PDE,
          E430028B21Rik
          -164.3kb Anti-sense Chr. 14 - 26664117 - 26669846 on NCBI Build GRCm38
          Downstream ankyrin repeat and SOCS box-containing 14 Asb14 Asb-14 58.3kb Sense Chr. 14 - 26894591 - 26916100 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P329178 AK016710.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11278159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11476451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11569372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11325319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11810195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11309410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11530426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29483727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432671_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432672_PM_at - -
          Mouse Genome 430 2.0 Array 1432671_PM_at - -
          Mouse Genome 430 2.0 Array 1432672_PM_at - -
          Mouse Genome 430 2.0 Array 1432672_at - -
          Mouse Genome 430 2.0 Array 1432671_at - -
          Mouse Expression Set 430 1432672_PM_at - -
          Mouse Expression Set 430 1432671_PM_at - -
          Mouse Expression Set 430 1432671_at - -
          Mouse Expression Set 430 1432672_at - -
          Affymetrix HT Mouse Genome 430B Array 1432672_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432671_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432672_at - -
          Affymetrix HT Mouse Genome 430B Array 1432671_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 432053 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0