Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03940793_s1
          See other MFAP3L GE Assays ›
          Gene Symbol:
          Mfap3l
          Gene Name:
          microfibrillar-associated protein 3-like
          Gene Aliases:
          4933428A15Rik, 5430405D20Rik, AI461995, AW125052, NYD-sp9, mKIAA0626
          Chromosome Location:
          Chr.8: 60632825 - 60676731 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03940793_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfap3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177881.1 NP_001171352.1 3 6259 83
          NM_001177882.1 NP_001171353.1 3 6027 83
          NM_001293769.1 NP_001280698.1 2 5721 83
          NM_027756.5 NP_082032.1 3 6151 83
          GenBank mRNA AK017269.1 - 1 1652 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF146299.1 - 1 Chr. 8 - 60676262 - 60676725 [-] on Build GRCm38 83
          GenBankmRNA BE196976.1 - 1 Chr. 8 - 60676267 - 60676726 [-] on Build GRCm38 83
          GenBankmRNA BB519927.1 - 1 112 Chr. 8 - 60676399 - 60676726 [+] on Build GRCm38 83
          GenBankmRNA CJ256615.1 - 1 307 Chr. 8 - 60676219 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA CJ253753.1 - 1 251 Chr. 8 - 60676260 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA BF322902.1 - 1 Chr. 8 - 60676257 - 60676729 [-] on Build GRCm38 83
          GenBankmRNA CJ254252.1 - 1 230 Chr. 8 - 60676281 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA CJ253659.1 - 1 270 Chr. 8 - 60676241 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA AI461995.1 - 1 Chr. 8 - 60676301 - 60676726 [-] on Build GRCm38 83
          GenBankmRNA CJ252103.1 - 1 268 Chr. 8 - 60676243 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA BG975950.1 - 1 253 Chr. 8 - 60676258 - 60676725 [+] on Build GRCm38 83
          GenBankmRNA BE133128.1 - 1 Chr. 8 - 60676291 - 60676729 [-] on Build GRCm38 83
          GenBankmRNA BE137600.1 - 1 341 Chr. 8 - 60676170 - 60676605 [+] on Build GRCm38 83
          GenBankmRNA AW985850.1 - 1 Chr. 8 - 60676306 - 60676726 [-] on Build GRCm38 83
          GenBankmRNA CJ251241.1 - 1 267 Chr. 8 - 60676244 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA CJ305925.1 - 1 188 Chr. 8 - 60676323 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA AW125052.1 - 1 Chr. 8 - 60676236 - 60676726 [-] on Build GRCm38 83
          GenBankmRNA BX515750.1 - 1 346 Chr. 8 - 60676174 - 60676713 [+] on Build GRCm38 83
          GenBankmRNA CJ258972.1 - 1 241 Chr. 8 - 60676270 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA CJ252403.1 - 1 280 Chr. 8 - 60676232 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA CJ301785.1 - 1 222 Chr. 8 - 60676289 - 60676727 [+] on Build GRCm38 83
          GenBankmRNA AI120312.1 - 1 341 Chr. 8 - 60676170 - 60676580 [+] on Build GRCm38 83
          GenBankmRNA BE650457.1 - 1 360 Chr. 8 - 60676151 - 60676726 [+] on Build GRCm38 83
          GenBankmRNA CJ255879.1 - 1 236 Chr. 8 - 60676275 - 60676727 [+] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream aminoadipate aminotransferase Aadat AI875679,
          Aadt,
          KATII,
          Kat2,
          mKat-2
          -129.2kb Sense Chr. 8 - 60505870 - 60545677 on NCBI Build GRCm38
          Downstream RIKEN cDNA 2700029M09 gene 2700029M09Rik C230006B22Rik 213.7kb Sense Chr. 8 - 60890393 - 60907582 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000646.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mfap3l

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 TC0800000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 NM_001177881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 NM_001177881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 NM_001177882.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 NM_001177882.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004314.mm.1 NM_027756.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 TC0800000646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 NM_001177881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 NM_001177881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 NM_001177882.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 NM_001177882.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004313.mm.1 NM_027756.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix