Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03940883_m1
          See other 5430425K12RIK GE Assays ›
          Gene Symbol:
          5430425K12Rik
          Gene Name:
          RIKEN cDNA 5430425K12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 80940403 - 80948597 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03940883_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430425K12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017339.1 - 1 - 2 246 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY717153.1 - 1 - 2 246 Chr. 13 - 80941059 - 80948576 [+] on Build GRCm38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream arrestin domain containing 3 Arrdc3 AI450344,
          mKIAA1376
          -45.0kb Sense Chr. 13 - 80883338 - 80896042 on NCBI Build GRCm38
          Downstream predicted gene 4211 Gm4211 9.1kb Anti-sense Chr. 13 - 80957664 - 80961434 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973115 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104884 AK017339.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07932062 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10655734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10439279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10655732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10985214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10733762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10565611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10925765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10729758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10595972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11113653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10890362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10565623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10591574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11016675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10593118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10595971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10925760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11088277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10890368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29173001 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0