Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03941385_s1
          See other SFPQ GE Assays ›
          Gene Symbol:
          Sfpq
          Gene Name:
          splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
          Gene Aliases:
          1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
          Chromosome Location:
          Chr.4: 127020560 - 127037016 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03941385_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfpq
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017658.1 - 1 151 94
          AK141302.1 - 1 1445 94
          BC033603.1 - 1 2843 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY717438.1 - 1 151 Chr. 4 - 127034571 - 127034950 [+] on Build GRCm38 94
          GenBankmRNA AU021761.1 - 1 Chr. 4 - 127034380 - 127034949 [-] on Build GRCm38 94
          GenBankmRNA BM934843.1 - 1 280 Chr. 4 - 127034443 - 127035091 [+] on Build GRCm38 94
          GenBankmRNA AU021830.1 - 1 Chr. 14 - 15098235 - 15098736 [+] on Build GRCm38 94
          GenBankmRNA BG061796.2 - 1 Chr. 4 - 127034459 - 127034949 [-] on Build GRCm38 94
          GenBankmRNA CX207240.1 - 1 237 Chr. 4 - 127034485 - 127034993 [+] on Build GRCm38 94
          GenBankmRNA BG068037.2 - 1 Chr. 4 - 127034381 - 127034949 [-] on Build GRCm38 94
          GenBankmRNA CD547993.1 - 1 Chr. 4 - 127034477 - 127034949 [-] on Build GRCm38 94
          GenBankmRNA AW554262.2 - 1 Chr. 4 - 127034478 - 127034949 [-] on Build GRCm38 94
          GenBankmRNA CD770415.1 - 1 114 Chr. 4 - 127034611 - 127035245 [+] on Build GRCm38 94
          GenBankmRNA AW910609.1 - 1 Chr. 4 - 127034482 - 127034952 [-] on Build GRCm38 94
          GenBankmRNA AU035904.1 - 1 437 Chr. 4 - 127034285 - 127035025 [+] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger,
          MYM-type 4
          Zmym4 6330503C17Rik,
          AI480785,
          AW493829,
          CDIR,
          D630001M21,
          MYM,
          Zfp262,
          Znf262,
          mKIAA0425
          -65.2kb Anti-sense Chr. 4 - 126861819 - 126969370 on NCBI Build GRCm38
          Downstream leucine rich repeat containing 3B Lrrc3b BC019794,
          LRP15
          258.8kb Anti-sense Chr. 14 - 15357515 - 15438987 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0