Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03941708_s1
          See other GM11346 GE Assays ›
          Gene Symbol:
          Gm11346
          Gene Name:
          X-linked lymphocyte-regulated 5 pseudogene
          Gene Aliases:
          5830431A10Rik, AI447617, Gm11347, OTTMUSG00000000601, OTTMUSG00000000602
          Chromosome Location:
          Chr.13: 24598124 - 24604767 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03941708_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024599.1 2 872 110
          GenBank mRNA AK017957.1 - 1 646 110
          BC064072.1 - 2 872 110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ236201.1 - 1 325 Chr. 13 - 24598030 - 24598693 [-] on Build GRCm38 110

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream family with sequence similarity 65,
          member B
          Fam65b Anti-sense Chr. 13 - 24501534 - 24733806 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700016G14 gene 1700016G14Rik 134.9kb Anti-sense Chr. 13 - 24733875 - 24746592 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001664.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 AK017957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 AK139092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 BC064072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 NR_024599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 NR_024599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 AK017957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 AK139092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394847 BC064072.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354066 AK033500.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354066 BC036986.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354066 M17192.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354066 NM_010455.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118813 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158814 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 AK017957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 AK139092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 BC064072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 NR_024599.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 AK017957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 AK139092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 BC064072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394847 NR_024599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 AK017957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 AK139092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 BC064072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 NR_024599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 AK017957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 AK139092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 BC064072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394847 NR_024599.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10649919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10677141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10649924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11022618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11022622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11022625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10738934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28887969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1436491_PM_at - -
          Mouse Genome 430 2.0 Array 1436491_at - -
          Mouse Genome 430 2.0 Array 1436491_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76024_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76024_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300010341.mm.1 TC1300001664.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010341.mm.1 NR_024599.1 Refseq
          Mouse Expression Set 430 1436491_at - -
          Mouse Expression Set 430 1436491_PM_at - -
          Murine Genome U74v2 Set 94136_PM_at - -
          Murine Genome U74v2 Set 94136_at - -
          Affymetrix HT Mouse Genome 430B Array 1436491_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436491_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 347436 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0