Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03941787_s1
          See other 5830437K03RIK GE Assays ›
          Gene Symbol:
          5830437K03Rik
          Gene Name:
          RIKEN cDNA 5830437K03 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03941787_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5830437K03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017976.1 - 1 1145 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ribosomal protein L34 Rpl34 1100001I22Rik -171.3kb Anti-sense Chr. 3 - 130726827 - 130730398 on NCBI Build GRCm38
          Downstream lymphoid enhancer binding factor 1 Lef1 3000002B05,
          AI451430,
          Lef-1
          207.2kb Sense Chr. 3 - 131110263 - 131224357 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2291405 AK017976.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02595217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02860885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02313112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03012947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22528847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02969214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02369785 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432731_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432732_PM_at - -
          Mouse Genome 430 2.0 Array 1432732_PM_at - -
          Mouse Genome 430 2.0 Array 1432731_PM_at - -
          Mouse Genome 430 2.0 Array 1432732_at - -
          Mouse Genome 430 2.0 Array 1432731_at - -
          Mouse Expression Set 430 1432732_PM_at - -
          Mouse Expression Set 430 1432731_at - -
          Mouse Expression Set 430 1432731_PM_at - -
          Mouse Expression Set 430 1432732_at - -
          Affymetrix HT Mouse Genome 430B Array 1432732_at - -
          Affymetrix HT Mouse Genome 430B Array 1432731_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432731_at - -
          Affymetrix HT Mouse Genome 430B Array 1432732_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 299125 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0