Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03942041_s1
          See other 6330405D24RIK GE Assays ›
          Gene Symbol:
          6330405D24Rik
          Gene Name:
          RIKEN cDNA 6330405D24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03942041_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6330405D24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018123.1 - 1 852 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream MAX gene associated Mga AV312082,
          C130042M01Rik,
          C80739,
          D030062C11Rik,
          Mad5
          Anti-sense Chr. 2 - 119871646 - 119970129 on NCBI Build GRCm38
          Downstream mitogen-activated protein kinase binding protein 1 Mapkbp1 2810483F24Rik,
          AW123212,
          JNKBP-1,
          Jnkbp1,
          mKIAA0596
          99.1kb Anti-sense Chr. 2 - 119972388 - 120027403 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150811 AK161433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150811 AK161433.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21571916 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10486321 - -
          Affymetrix HT MG-430 PM Array Plate 1431346_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10486320 - -
          Mouse Genome 430 2.0 Array 1431346_at - -
          Mouse Genome 430 2.0 Array 1431346_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70715_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70715_at - -
          Mouse Expression Set 430 1431346_at - -
          Mouse Expression Set 430 1431346_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431346_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431346_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104050315 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2435825 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237795 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0