Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03942422_s1
          See other 6530406A20RIK GE Assays ›
          Gene Symbol:
          6530406A20Rik
          Gene Name:
          RIKEN cDNA 6530406A20 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03942422_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6530406A20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AA739088.1 - 1 114 90
          AK018324.1 - 1 847 90
          AK087211.1 - 1 2340 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM502661.1 - 1 Chr. 12 - 112955631 - 112956147 [+] on Build GRCm38 90
          GenBankmRNA BQ946513.1 - 1 420 Chr. 12 - 112955640 - 112956479 [-] on Build GRCm38 90
          GenBankmRNA BB534254.2 - 1 130 Chr. 12 - 112955547 - 112956192 [-] on Build GRCm38 90
          GenBankmRNA BM506214.1 - 1 Chr. 12 - 112955790 - 112956260 [-] on Build GRCm38 90
          GenBankmRNA CX206385.1 - 1 498 Chr. 12 - 112955637 - 112956557 [-] on Build GRCm38 90
          GenBankmRNA AW244274.1 - 1 Chr. 12 - 112955875 - 112956158 [+] on Build GRCm38 90
          GenBankmRNA BQ042583.1 - 1 391 Chr. 12 - 112955661 - 112956450 [-] on Build GRCm38 90
          GenBankmRNA CD803645.1 - 1 398 Chr. 12 - 112955826 - 112956457 [-] on Build GRCm38 90
          GenBankmRNA CK780393.1 - 1 398 Chr. 12 - 112955826 - 112956457 [-] on Build GRCm38 90
          GenBankmRNA BM945138.1 - 1 391 Chr. 12 - 112955711 - 112956450 [-] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream BRF1 homolog,
          subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae)
          Brf1 2510002F24Rik,
          GTF3B,
          TAF3C,
          TAFIII90,
          TFIIIB90,
          mTFIIIB90
          Sense Chr. 12 - 112955631 - 113000707 on NCBI Build GRCm38
          Downstream BTB (POZ) domain containing 6 Btbd6 18.1kb Anti-sense Chr. 12 - 112974998 - 112979493 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548711 - -
          Whole Mouse Genome G4122A A_51_P249516 AK018324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249516 AK087211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249516 AA739088.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115140 - -
          Agilent Mouse cDNA Microarray (G4104A) 1226854 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429153_PM_at - -
          Mouse Genome 430 2.0 Array 1429153_PM_at - -
          Mouse Genome 430 2.0 Array 1429153_at - -
          Mouse Expression Set 430 1429153_PM_at - -
          Mouse Expression Set 430 1429153_at - -
          Affymetrix HT Mouse Genome 430B Array 1429153_at - -
          Affymetrix HT Mouse Genome 430B Array 1429153_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 556731 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104810450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix