Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03942732_s1
          See other 9030407C09RIK GE Assays ›
          Gene Symbol:
          9030407C09Rik
          Gene Name:
          RIKEN cDNA 9030407C09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03942732_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9030407C09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018489.1 - 1 956 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ADP-ribosylation factor-like 6 interacting protein 6 Arl6ip6 2310057C01Rik,
          2610529A11Rik,
          Aip-6
          -658.6kb Sense Chr. 2 - 53192084 - 53219221 on NCBI Build GRCm38
          Downstream reprimo,
          TP53 dependent G2 arrest mediator candidate
          Rprm 2410012A13Rik,
          Reprimo
          204.6kb Anti-sense Chr. 2 - 54084093 - 54085552 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_52_P817958 AK018489.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P02030792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02030797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21226620 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432691_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432690_PM_at - -
          Mouse Genome 430 2.0 Array 1432690_PM_at - -
          Mouse Genome 430 2.0 Array 1432691_at - -
          Mouse Genome 430 2.0 Array 1432690_at - -
          Mouse Genome 430 2.0 Array 1432691_PM_at - -
          Mouse Expression Set 430 1432691_at - -
          Mouse Expression Set 430 1432690_at - -
          Mouse Expression Set 430 1432690_PM_at - -
          Mouse Expression Set 430 1432691_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432691_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432690_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432690_at - -
          Affymetrix HT Mouse Genome 430B Array 1432691_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0