Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03942903_s1
          See other 9130002K18RIK GE Assays ›
          Gene Symbol:
          9130002K18Rik
          Gene Name:
          RIKEN cDNA 9130002K18 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03942903_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9130002K18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018586.1 - 1 2255 140

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 875 Mir875 Mirn875,
          mir-875,
          mmu-mir-875
          -126.1kb Anti-sense Chr. 15 - 35660970 - 35661047 on NCBI Build GRCm38
          Downstream cytochrome c oxidase subunit VIc Cox6c 141.7kb Anti-sense Chr. 15 - 35931976 - 35938246 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2255944 AK018586.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115036 - -
          Whole Mouse Genome G4122A A_51_P258859 AK018586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124978 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12165312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11991598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12165311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11991576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30135052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431525_PM_at - -
          Mouse Genome 430 2.0 Array 1431525_PM_at - -
          Mouse Genome 430 2.0 Array 1431525_at - -
          Mouse Expression Set 430 1431525_at - -
          Mouse Expression Set 430 1431525_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431525_at - -
          Affymetrix HT Mouse Genome 430B Array 1431525_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100770133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0