Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03944869_s1
          See other 6230400D17RIK GE Assays ›
          Gene Symbol:
          6230400D17Rik
          Gene Name:
          RIKEN cDNA 6230400D17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 20702012 - 20703099 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03944869_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6230400D17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029446.1 1 916 101
          GenBank mRNA AK078695.1 - 1 849 101
          AK156493.1 - 828 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA533466.1 - 1 314 Chr. 14 - 20701950 - 20702497 [-] on Build GRCm38 101
          GenBankmRNA BB064773.2 - 1 507 Chr. 14 - 20702012 - 20702681 [-] on Build GRCm38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream coiled-coil-helix-coiled-coil-helix domain containing 1 Chchd1 1110001O19Rik,
          2400010G13Rik,
          MRP-S37
          Anti-sense Chr. 14 - 20703027 - 20704425 on NCBI Build GRCm38
          Downstream zinc finger SWIM-type containing 8 Zswim8 2310021P13Rik,
          4832404P21Rik,
          Kiaa0913,
          mKIAA0913
          4.3kb Anti-sense Chr. 14 - 20707319 - 20723619 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001548.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2324360 AK018074.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143951 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2324360 AK018074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301173 AK018074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P242371 AK078695.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P242371 AK156493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P242371 NR_029446.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10417949 - -
          Affymetrix HT MG-430 PM Array Plate 1460473_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10417946 - -
          Mouse Genome 430 2.0 Array 1460473_PM_at - -
          Mouse Genome 430 2.0 Array 1460473_at - -
          Affymetrix Mouse Clariom S Array PSR1400011599.mm.1 TC1400001548.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011599.mm.1 NR_029446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011599.mm.1 NR_029446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011602.mm.1 TC1400001548.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011602.mm.1 NR_029446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011602.mm.1 NR_029446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011598.mm.1 TC1400001548.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011598.mm.1 NR_029446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011598.mm.1 NR_029446.1 Refseq
          Mouse Expression Set 430 1460473_at - -
          Mouse Expression Set 430 1460473_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460473_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460473_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106980162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0