Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03945276_m1
          See other A530065N20RIK GE Assays ›
          Gene Symbol:
          A530065N20Rik
          Gene Name:
          RIKEN cDNA A530046M15 gene
          Gene Aliases:
          A530065N20
          Chromosome Location:
          Chr.13: 60029711 - 60116669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03945276_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530065N20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046142.1 2 - 3 341 68
          GenBank mRNA AK080130.1 - 2 - 3 342 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB637326.1 - 2 - 3 342 Chr. 13 - 60029711 - 60113578 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger,
          CCHC domain containing 6
          Zcchc6 6030448M23Rik,
          AA420405
          -287.3kb Anti-sense Chr. 13 - 59771876 - 59823167 on NCBI Build GRCm38
          Downstream growth arrest specific 1 Gas1 AW554192,
          Gas-1
          57.7kb Anti-sense Chr. 13 - 60174405 - 60177535 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000730.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2404394 AK080130.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16486 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10934144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11069191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10889965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11041390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10703569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11069188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10889415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10610153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10556231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10702063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10934139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10995123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11021911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29083658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10702061 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300004859.mm.1 TC1300000730.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004859.mm.1 NR_046142.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004859.mm.1 NR_046142.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 304005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0