Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03945537_s1
          See other A730060N03RIK GE Assays ›
          Gene Symbol:
          A730060N03Rik
          Gene Name:
          RIKEN cDNA A730060N03 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03945537_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730060N03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK081056.1 - 1 2354 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream calcium channel,
          voltage-dependent,
          gamma subunit 2
          Cacng2 AW060990,
          B230105C07Rik,
          B930041E13Rik,
          stargazer,
          stargazin,
          stg,
          wag,
          waggler
          -21.2kb Anti-sense Chr. 15 - 77993623 - 78119280 on NCBI Build GRCm38
          Downstream intraflagellar transport 27 Ift27 2600013G09Rik,
          Rabl4
          15.4kb Anti-sense Chr. 15 - 78159463 - 78174108 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141419 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03076471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147460 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109338 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12371837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12125901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12354608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12244973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12371849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12371861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11964429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12125905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30358046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30357965 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887706 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101400162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0