Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03946545_m1
          See other D330046F09RIK GE Assays ›
          Gene Symbol:
          D330046F09Rik
          Gene Name:
          RIKEN cDNA D330046F09 gene
          Gene Aliases:
          Gm808
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03946545_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D330046F09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK084812.1 - 3 - 4 209 99
          BB659569.1 - 3 - 4 209 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4933425B07 gene 4933425B07Rik -8.4kb Sense Chr. 14 - 46602806 - 46637423 on NCBI Build GRCm38
          Downstream cyclin-dependent kinase inhibitor 3 Cdkn3 2410006H10Rik,
          KAP
          109.9kb Anti-sense Chr. 14 - 46760464 - 46771529 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AF326732.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AK010351.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AK012011.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AK089939.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AK132467.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 AK162249.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 BC020026.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 NM_023284.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765925 XM_001479748.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133209 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159888 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159888 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08100360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08100387 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11260117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11231420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11583498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11670588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11384895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11566644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11473422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11437898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11702607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11566630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11439574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11384896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11234970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11544639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11384900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29577674 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1446825_at - -
          Mouse Genome 430 2.0 Array 1446825_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6200647 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0