Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03946713_s1
          See other GM9938 GE Assays ›
          Gene Symbol:
          Gm9938
          Gene Name:
          predicted gene 9938
          Gene Aliases:
          ENSMUSG00000054178
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03946713_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm9938
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK085249.1 - 1 1347 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM937180.1 - 1 Chr. 19 - 23724184 - 23724855 [-] on Build GRCm38 79
          GenBankmRNA CV555887.1 - 1 192 Chr. 19 - 23724433 - 23725064 [+] on Build GRCm38 79
          GenBankmRNA CV555924.1 - 1 165 Chr. 19 - 23724460 - 23725055 [+] on Build GRCm38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein prenyltransferase alpha subunit repeat containing 1 Ptar1 1700084D09Rik,
          2010013L17Rik,
          4930428J16Rik,
          AI449677
          -187.0 Sense Chr. 19 - 23686898 - 23723092 on NCBI Build GRCm38
          Downstream amyloid beta (A4) precursor protein binding,
          family A,
          member 1
          Apba1 6430513E09Rik,
          Lin-10,
          Mint,
          Mint1,
          Urop11,
          X11,
          X11alpha
          33.8kb Sense Chr. 19 - 23758876 - 23949598 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P956363 AK085249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P956363 AK085249.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171589 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P956363 AK085249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P956363 AK085249.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171589 - -
          Whole Mouse Genome G4122A A_52_P956363 AK085249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P956363 AK085249.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129659 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14485813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32019357 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104210673 - -
          Illumina mouse-6 v1.0 expression beadchip 104210673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219094 - -
          Illumina Sentrix mouse-6.v1 bead array 104210673 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104210673 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219094 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0