Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03947214_m1
          See other GM6980 GE Assays ›
          Gene Symbol:
          Gm6980
          Gene Name:
          predicted gene 6980
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 51915912 - 51930268 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03947214_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6980
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK086943.1 - 394 75
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB605849.2 - 4 - 5 393 Chr. 9 - 51913800 - 51931661 [-] on Build GRCm38 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Rho GTPase activating protein 20,
          opposite strand
          Arhgap20os 9230115E21Rik -37.9kb Sense Chr. 9 - 51839493 - 51875885 on NCBI Build GRCm38
          Downstream ferredoxin 1 Fdx1 27.1kb Sense Chr. 9 - 51943023 - 51963602 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21815 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125093 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956495 AK086943.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125093 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173562 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132693 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173562 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07595306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07895427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07924847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07497253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08197177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07598377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07895428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08034651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07595301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07958634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08197172 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107050746 - -
          Illumina mouse-6 v1.0 expression beadchip 107050746 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215523 - -
          Illumina Sentrix mouse-6.v1 bead array 107050746 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107050746 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215523 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0