Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03947390_m1
          See other E330023G01RIK GE Assays ›
          Gene Symbol:
          E330023G01Rik
          Gene Name:
          RIKEN cDNA E330023G01 gene
          Gene Aliases:
          E330023G01
          Chromosome Location:
          Chr.9: 98748599 - 98820087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03947390_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E330023G01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045332.1 2 - 3 281 72
          GenBank mRNA AK087804.1 - 2 - 3 282 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB666609.1 - 2 - 3 282 Chr. 9 - 98749451 - 98820087 [-] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitochondrial ribosomal protein S22 Mrps22 3100002P07Rik,
          Rpms22
          -146.9kb Sense Chr. 9 - 98588730 - 98601679 on NCBI Build GRCm38
          Downstream proline rich 23A Prr23a EG623166,
          Gm6400
          84.3kb Anti-sense Chr. 9 - 98842587 - 98843645 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002912.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289287 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK012030.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK014459.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK027927.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK076203.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK083347.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK137039.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 AK156414.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 BC027269.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 NM_025703.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550131 XM_001473537.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 AK009373.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 AK035493.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 AK148986.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 AK173265.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 BC032179.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 BC051093.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 BC076619.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928888 NM_001081058.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51442 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142739 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142739 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06896236 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06896292 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06896329 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06896292 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900023970.mm.1 TC0900002912.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023970.mm.1 NR_045332.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023970.mm.1 NR_045332.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 630943 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100510048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-56f585df74-hnff6:80/100.66.130.62:80.
          git-commit: b2a1b94c9c7f5a2900588881f383e993a09fadb0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0