Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500009950.mm.1 |
TC1500001253.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009950.mm.1 |
NR_045711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009950.mm.1 |
NR_045711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009952.mm.1 |
TC1500001253.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009952.mm.1 |
NR_045711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009952.mm.1 |
NR_045711.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009949.mm.1 |
TC1500001253.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500009949.mm.1 |
NR_045711.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500009949.mm.1 |
NR_045711.1 |
Refseq |