Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948586_m1
          See other GM2516 GE Assays ›
          Gene Symbol:
          Gm2516
          Gene Name:
          predicted gene 2516
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 50368112 - 50416089 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948586_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2516
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046067.1 5 - 6 746 94
          GenBank mRNA AK135701.1 - 5 - 6 750 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream teneurin transmembrane protein 3 Tenm3 2610100B16Rik,
          Odz1,
          Odz3,
          Ten-m3,
          mKIAA1455
          -1.5mb Anti-sense Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38
          Downstream predicted gene,
          32975
          Gm32975 761.6kb Sense Chr. 8 - 51177683 - 51180608 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000575.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074723 AK135701.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK028084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK029567.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK040643.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK045288.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK051345.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK054550.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK076989.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK085626.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK136187.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK168431.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 AK169780.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 BC085321.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753672 NM_178665.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89576 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074723 AK135701.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01735660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06509641 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06509740 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06509684 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06509740 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06509684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25859857 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800004025.mm.1 TC0800000575.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004025.mm.1 NR_046067.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004025.mm.1 NR_046067.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0