Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948650_m1
          See other GM626 GE Assays ›
          Gene Symbol:
          Gm626
          Gene Name:
          predicted gene 626
          Gene Aliases:
          ENSMUSG00000071536, Frmpd2, Gm1582
          Chromosome Location:
          Chr.14: 33471696 - 33575269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948650_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm626
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK136780.1 - 3 - 4 1579 60
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG404624.1 - 1 - 2 340 Chr. 14 - 33571677 - 33574691 [+] on Build GRCm38 60

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mitogen-activated protein kinase 8 Mapk8 AI849689,
          JNK,
          JNK1,
          Prkm8,
          SAPK1
          -123.6kb Anti-sense Chr. 14 - 33377898 - 33447214 on NCBI Build GRCm38
          Downstream protein tyrosine phosphatase,
          non-receptor type 20
          Ptpn20 typ 13.9kb Sense Chr. 14 - 33589201 - 33640754 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094173 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094172 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094173 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413992 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413998 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413984 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413993 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413981 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413999 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413987 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413997 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414003 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413983 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413985 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413977 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413989 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413995 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414001 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413991 - -
          Affymetrix Mouse Gene 1.1 ST Array 10413979 - -
          Mouse Expression Set 430 1458506_at - -
          Mouse Expression Set 430 1458506_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0