Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948899_m1
          See other A230045G11RIK GE Assays ›
          Gene Symbol:
          A230045G11Rik
          Gene Name:
          RIKEN cDNA A230045G11 gene
          Gene Aliases:
          EG624086
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948899_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230045G11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK133970.1 - 2 - 3 370 94
          AK138253.1 - 2 - 3 326 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU923922.1 - 1 - 2 548 Chr. 17 - 3113293 - 3114732 [-] on Build GRCm38 94
          GenBankmRNA BF451515.1 - 2 - 3 332 Chr. 17 - 3112699 - 3114757 [-] on Build GRCm38 94
          GenBankmRNA BE945457.1 - 2 - 3 461 Chr. 17 - 3112529 - 3114885 [-] on Build GRCm38 94
          GenBankmRNA AV511959.1 - 2 - 3 334 Chr. 17 - 3112521 - 3114760 [-] on Build GRCm38 94
          GenBankmRNA CJ170071.1 - 2 - 3 358 Chr. 17 - 3113292 - 3114783 [-] on Build GRCm38 94
          GenBankmRNA BF472402.1 - 1 - 2 Chr. 17 - 3113294 - 3114878 [+] on Build GRCm38 94
          GenBankmRNA BY723858.1 - 2 - 3 324 Chr. 17 - 3104197 - 3114749 [-] on Build GRCm38 94
          GenBankmRNA BY239044.1 - 2 - 3 223 Chr. 17 - 3104352 - 3114648 [-] on Build GRCm38 94
          GenBankmRNA BG802605.1 - 2 - 3 332 Chr. 17 - 3112362 - 3114757 [-] on Build GRCm38 94
          GenBankmRNA BY239016.1 - 2 - 3 382 Chr. 17 - 3113295 - 3114805 [-] on Build GRCm38 94
          GenBankmRNA BY252268.1 - 2 - 3 223 Chr. 17 - 3104325 - 3114648 [-] on Build GRCm38 94
          GenBankmRNA CJ048768.1 - 2 - 3 370 Chr. 17 - 3112364 - 3114795 [-] on Build GRCm38 94
          GenBankmRNA CF731830.1 - 2 - 3 331 Chr. 17 - 3112313 - 3114756 [-] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylserine decarboxylase,
          pseudogene 2
          Pisd-ps2 D030013I16Rik -29.1kb Sense Chr. 17 - 3064318 - 3084183 on NCBI Build GRCm38
          Downstream SR-related CTD-associated factor 8 Scaf8 A630086M08Rik,
          A930036P18Rik,
          AA408306,
          AI447644,
          AI448652,
          AU015035,
          Rbm16,
          mKIAA1116
          627.0 Anti-sense Chr. 17 - 3114972 - 3198859 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066897 AK133970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066897 AK138253.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106295 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066897 AK133970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066897 AK138253.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03267810 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03267807 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03267807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13483520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31003180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13483509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13158604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13158609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13417113 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0