Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949078_s1
          See other 5430434G16RIK GE Assays ›
          Gene Symbol:
          5430434G16Rik
          Gene Name:
          RIKEN cDNA 5430434G16 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949078_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430434G16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK139147.1 - 1 2052 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG084096.2 - 1 384 Chr. 15 - 36250419 - 36251089 [-] on Build GRCm38 90
          GenBankmRNA AU040386.1 - 1 Chr. 15 - 36250390 - 36250973 [+] on Build GRCm38 90
          GenBankmRNA BE991385.1 - 1 Chr. 15 - 36250491 - 36250912 [+] on Build GRCm38 90
          GenBankmRNA AA166103.1 - 1 Chr. 15 - 36250385 - 36250809 [+] on Build GRCm38 90
          GenBankmRNA BM936045.1 - 1 223 Chr. 15 - 36250490 - 36250912 [-] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 8097 Mir8097 Gm27497,
          mmu-mir-8097
          -10.3kb Sense Chr. 15 - 36240046 - 36240164 on NCBI Build GRCm38
          Downstream ankyrin repeat domain 46 Ankrd46 1110054N06Rik,
          AI314978,
          AI987733
          224.9kb Sense Chr. 15 - 36477668 - 36503426 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2262287 AK139147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1034794 AK139147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1034794 AK017390.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454383_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441976_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432850_PM_at - -
          Mouse Genome 430 2.0 Array 1432850_PM_at - -
          Mouse Genome 430 2.0 Array 1454383_at - -
          Mouse Genome 430 2.0 Array 1441976_at - -
          Mouse Genome 430 2.0 Array 1441976_PM_at - -
          Mouse Genome 430 2.0 Array 1432850_at - -
          Mouse Genome 430 2.0 Array 1454383_PM_at - -
          Mouse Expression Set 430 1432850_PM_at - -
          Mouse Expression Set 430 1432850_at - -
          Mouse Expression Set 430 1441976_PM_at - -
          Mouse Expression Set 430 1454383_PM_at - -
          Mouse Expression Set 430 1454383_at - -
          Mouse Expression Set 430 1441976_at - -
          Affymetrix HT Mouse Genome 430B Array 1432850_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441976_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432850_at - -
          Affymetrix HT Mouse Genome 430B Array 1441976_at - -
          Affymetrix HT Mouse Genome 430B Array 1454383_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454383_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 669641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5df8fb85f4-n94h7:80/100.66.130.62:80.
          git-commit: 8f66f361c5203ebe605e386d8a840ec15b9b8304
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0