Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949341_s1
          See other MALAT1 GE Assays ›
          Gene Symbol:
          Malat1
          Gene Name:
          metastasis associated lung adenocarcinoma transcript 1 (non-coding RNA)
          Gene Aliases:
          2210401K01Rik, 9430072K23Rik, AI647968, Neat2
          Chromosome Location:
          Chr.19: 5795690 - 5802671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949341_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Malat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002847.2 1 6367 86
          GenBank mRNA AK141413.1 - 1 1451 86
          EF177380.1 - 1 6367 86
          FJ209304.1 - 1 6367 86
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY642518.1 - 1 175 Chr. 19 - 5796168 - 5796479 [-] on Build GRCm38 86
          GenBankmRNA BU513766.1 - 1 582 Chr. 19 - 5795934 - 5796887 [-] on Build GRCm38 85
          GenBankmRNA AW210565.1 - 1 Chr. 19 - 5796055 - 5796414 [+] on Build GRCm38 86
          GenBankmRNA AY722410.1 - 1 6367 Chr. - - [-] on Build 86
          GenBankmRNA BU697194.1 - 1 257 Chr. 19 - 5796056 - 5796561 [-] on Build GRCm38 86
          GenBankmRNA CD774784.1 - 1 Chr. 19 - 5795982 - 5796639 [+] on Build GRCm38 86
          GenBankmRNA CA587592.1 - 1 Chr. 19 - 5796169 - 5796420 [+] on Build GRCm38 86
          GenBankmRNA DV065135.1 - 1 545 Chr. 19 - 5795980 - 5796808 [-] on Build GRCm38 86
          GenBankmRNA AI647968.1 - 1 Chr. 19 - 5795999 - 5796555 [+] on Build GRCm38 86
          GenBankmRNA BY616255.1 - 1 218 Chr. 19 - 5796168 - 5796520 [-] on Build GRCm38 86
          GenBankmRNA AU260380.1 - 1 118 Chr. 19 - 5796118 - 5796422 [-] on Build GRCm38 86
          GenBankmRNA DV063405.1 - 446 Chr. - - [-] on Build 86
          GenBankmRNA BB418323.2 - 1 359 Chr. 19 - 5796000 - 5796654 [-] on Build GRCm38 86
          GenBankmRNA DV065019.1 - 1 545 Chr. 19 - 5795980 - 5796808 [-] on Build GRCm38 86
          GenBankmRNA BU921384.1 - 1 167 Chr. 19 - 5795982 - 5796471 [-] on Build GRCm38 86
          GenBankmRNA BP426953.1 - 1 188 Chr. 19 - 5796000 - 5796492 [-] on Build GRCm38 86
          GenBankmRNA AW824837.1 - 1 357 Chr. 19 - 5796259 - 5796662 [-] on Build GRCm38 86
          GenBankmRNA BG806067.1 - 1 456 Chr. 19 - 5796162 - 5796760 [-] on Build GRCm38 86
          GenBankmRNA BU520506.1 - 1 462 Chr. 19 - 5795817 - 5796765 [-] on Build GRCm38 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SCY1-like 1 (S. cerevisiae) Scyl1 2810011O19Rik,
          C85140,
          Ntkl,
          mdf,
          mfd,
          p105
          -24.6kb Sense Chr. 19 - 5758427 - 5771414 on NCBI Build GRCm38
          Downstream nuclear paraspeckle assembly transcript 1 (non-protein coding) Neat1 2310043N10Rik,
          VINC
          44.5kb Sense Chr. 19 - 5842302 - 5845478 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000984.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          nuclear speck organization
          regulation of synapse organization
          positive regulation of myoblast proliferation
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900008901.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008901.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008900.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008900.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008899.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008899.mm.1 NR_002847.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0