Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949830_m1
          See other 4930405D11RIK GE Assays ›
          Gene Symbol:
          4930405D11Rik
          Gene Name:
          RIKEN cDNA 4930405D11 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 90804691 - 90895152 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949830_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930405D11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045956.1 2 - 3 190 92
          GenBank mRNA AK147079.1 - 2 - 3 191 92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY740750.1 - 2 - 3 189 Chr. 11 - 90804692 - 90895149 [+] on Build GRCm38 92
          GenBankmRNA BY761038.1 - 2 - 3 195 Chr. 11 - 90804691 - 90895152 [+] on Build GRCm38 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream target of myb1-like 1 (chicken) Tom1l1 2310045L10Rik,
          C80573,
          Srcasm
          -135.4kb Anti-sense Chr. 11 - 90645690 - 90687661 on NCBI Build GRCm38
          Downstream kinesin family member 2B Kif2b 1700063D03Rik 680.1kb Anti-sense Chr. 11 - 91575286 - 91577555 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001402.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 182813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876600 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 116065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134042 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07439802 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07439725 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02393238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02393215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07439760 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07439642 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07439642 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02393238 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07439760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09375726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09028208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08979121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09375094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09432594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08979124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09029252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08979126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09375095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09206330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09480523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09444185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09373662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09669800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09202122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09029251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09017956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09223858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09582106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09562264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09439585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09017959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09588081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09534675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09375093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09481013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09640878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09110477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09640877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09592213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09455402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09631374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09110469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28013981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28014411 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100013099.mm.1 TC1100001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013099.mm.1 NR_045956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013099.mm.1 NR_045956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013100.mm.1 TC1100001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013100.mm.1 NR_045956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013100.mm.1 NR_045956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013110.mm.1 TC1100001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013110.mm.1 NR_045956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013110.mm.1 NR_045956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013112.mm.1 TC1100001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013112.mm.1 NR_045956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013112.mm.1 NR_045956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013109.mm.1 TC1100001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013109.mm.1 NR_045956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013109.mm.1 NR_045956.1 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104540504 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0