Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03950059_s1
          See other KDM5C GE Assays ›
          Gene Symbol:
          Kdm5c
          Gene Name:
          lysine (K)-specific demethylase 5C
          Gene Aliases:
          D930009K15Rik, Jarid1c, Smcx, mKIAA0234
          Chromosome Location:
          Chr.X: 152233196 - 152279102 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03950059_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdm5c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013668.4 NP_038696.2 26 8792 71
          GenBank mRNA AK148627.1 - 1 452 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF901759.1 - 1 417 Chr. X - 152276460 - 152277061 [+] on Build GRCm38 71
          GenBankmRNA CA529895.1 - 1 442 Chr. X - 152276435 - 152277033 [+] on Build GRCm38 71
          GenBankmRNA CN671816.1 - 1 443 Chr. X - 152276434 - 152276931 [+] on Build GRCm38 71
          GenBankmRNA BB623800.1 - 1 452 Chr. X - 152276426 - 152277040 [+] on Build GRCm38 71
          GenBankmRNA BG298682.1 - 192 Chr. - - [-] on Build 71
          GenBankmRNA BM947464.1 - 1 77 Chr. X - 152276800 - 152277382 [+] on Build GRCm38 71
          GenBankmRNA CN531744.1 - 1 441 Chr. X - 152276436 - 152277044 [+] on Build GRCm38 71
          GenBankmRNA CA895842.1 - 1 442 Chr. X - 152276435 - 152277014 [+] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream IQ motif and Sec7 domain 2 Iqsec2 Brag1,
          mKIAA0522
          -51.2kb Sense Chr. X - 152144183 - 152225237 on NCBI Build GRCm38
          Downstream Kdm5c adjacent non-coding transcript Kantr 2810454L23Rik,
          2900056M20Rik,
          Spasm
          15.7kb Anti-sense Chr. X - 152294824 - 152327493 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001541.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kdm5c

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 demethylation
          regulation of circadian rhythm
          negative regulation of transcription, DNA-templated
          rhythmic process
          sulfonate dioxygenase activity
          DNA binding
          zinc ion binding
          oxidoreductase activity
          2-oxoglutarate-dependent dioxygenase activity
          2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
          procollagen-proline dioxygenase activity
          histone H3-methyl-lysine-4 demethylase activity
          hypophosphite dioxygenase activity
          identical protein binding
          DNA-N1-methyladenine dioxygenase activity
          metal ion binding
          dioxygenase activity
          C-19 gibberellin 2-beta-dioxygenase activity
          C-20 gibberellin 2-beta-dioxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00009821.mm.1 TC0X00001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009821.mm.1 NM_013668.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009824.mm.1 TC0X00001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009824.mm.1 NM_013668.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009820.mm.1 TC0X00001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009820.mm.1 NM_013668.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009819.mm.1 TC0X00001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009819.mm.1 NM_013668.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0