Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952353_m1
          See other 4930471G03RIK GE Assays ›
          Gene Symbol:
          4930471G03Rik
          Gene Name:
          RIKEN cDNA 4930471G03 gene
          Gene Aliases:
          1700007N01Rik
          Chromosome Location:
          Chr.18: 36106203 - 36109954 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952353_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930471G03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015571.1 1 - 2 99 99
          GenBank mRNA AK015550.1 - 1 - 2 109 99
          BC048509.1 - 1 - 2 97 99
          BC061192.1 - 1 - 2 85 99
          BC100467.1 - 1 - 2 99 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI615320.1 - 1 - 2 106 Chr. 18 - 36106196 - 36109610 [+] on Build GRCm38 99
          GenBankmRNA CA466132.1 - 1 - 2 100 Chr. 18 - 36106205 - 36109954 [+] on Build GRCm38 99
          GenBankmRNA BY715098.1 - 109 Chr. - - [-] on Build 99
          GenBankmRNA CN843815.1 - 1 - 2 102 Chr. 18 - 36106203 - 36109952 [+] on Build GRCm38 99
          GenBankmRNA BU938417.1 - 80 Chr. - - [-] on Build 99
          GenBankmRNA BU935831.1 - 88 Chr. - - [-] on Build 99
          GenBankmRNA AA492618.1 - 1 - 2 106 Chr. 18 - 36106196 - 36109526 [+] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream neuregulin 2 Nrg2 Don1,
          NTAK
          Anti-sense Chr. 18 - 36016758 - 36199047 on NCBI Build GRCm38
          Downstream purine rich element binding protein A Pura CAGER-1,
          Pur-alpha,
          ssCRE-BP
          170.6kb Sense Chr. 18 - 36280551 - 36291721 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000323.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 514017 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03450094 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14096744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13621500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14019258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13632411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14019267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14072587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13622118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14072590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14037921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13812744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14054975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13739586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14072585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13734000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14019259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13782871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13692842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13642409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13692841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13632421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14024577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13937606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13967207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31622082 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10454876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454874 - -
          Affymetrix HT MG-430 PM Array Plate 1453838_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10454873 - -
          Mouse Genome 430 2.0 Array 1453838_PM_at - -
          Mouse Genome 430 2.0 Array 1453838_at - -
          Affymetrix Mouse Clariom S Array PSR1800002414.mm.1 TC1800000323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002414.mm.1 NR_015571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002414.mm.1 NR_015571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002412.mm.1 TC1800000323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002412.mm.1 NR_015571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002412.mm.1 NR_015571.1 Refseq
          Affymetrix HT Mouse Genome 430B Array 1453838_at - -
          Affymetrix HT Mouse Genome 430B Array 1453838_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 845473 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225593 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0