Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952492_s1
          See other 2310061G22RIK GE Assays ›
          Gene Symbol:
          2310061G22Rik
          Gene Name:
          RIKEN cDNA 2310061G22 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952492_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310061G22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK009999.1 - 1 1053 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eyes absent 2 homolog (Drosophila) Eya2 Eab1 Sense Chr. 2 - 165595028 - 165771727 on NCBI Build GRCm38
          Downstream zinc finger,
          MYND-type containing 8
          Zmynd8 1110013E22Rik,
          2010005I16Rik,
          3632413B07Rik,
          AI316811,
          AL024039,
          Prkcbp1,
          RACK7
          169.7kb Anti-sense Chr. 2 - 165784152 - 165898984 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02201963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02192530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02029190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01373506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01835409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01305377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01373508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01305376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02265450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01442692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01305373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01373504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02115782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01647566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01305365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02115783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21824911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02265449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430493_PM_at - -
          Mouse Genome 430 2.0 Array 1430493_PM_at - -
          Mouse Genome 430 2.0 Array 1430493_at - -
          Mouse Expression Set 430 1430493_PM_at - -
          Mouse Expression Set 430 1430493_at - -
          Affymetrix HT Mouse Genome 430B Array 1430493_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430493_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0