Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952602_g1
          See other 2410012E07RIK GE Assays ›
          Gene Symbol:
          2410012E07Rik
          Gene Name:
          RIKEN cDNA 2410012E07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 70852933 - 70873954 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952602_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2410012E07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045939.1 3 - 4 347 71
          GenBank mRNA AK010467.1 - 3 - 4 348 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream docking protein 2 Dok2 DokR,
          Frip
          -93.8kb Sense Chr. 14 - 70764528 - 70778494 on NCBI Build GRCm38
          Downstream glial cell line derived neurotrophic factor family receptor alpha 2 Gfra2 16.2kb Sense Chr. 14 - 70890107 - 70979840 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001011.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2311258 AK010467.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799510 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127706 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02880057 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108513 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11381277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11460127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11708815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11442553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11554704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11171990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11381273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11233269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11815858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11135217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11744209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11334754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11381274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11794428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11554697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11233264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11457275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29703888 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432118_PM_at - -
          Mouse Genome 430 2.0 Array 1432118_at - -
          Mouse Genome 430 2.0 Array 1432118_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400008119.mm.1 TC1400001011.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008119.mm.1 NR_045939.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008119.mm.1 NR_045939.1 Refseq
          Mouse Expression Set 430 1432118_at - -
          Mouse Expression Set 430 1432118_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432118_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432118_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0