Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952635_g1
          See other MAN2C1OS GE Assays ›
          Gene Symbol:
          Man2c1os
          Gene Name:
          mannosidase, alpha, class 2C, member 1, opposite strand
          Gene Aliases:
          2410133F24Rik, AI851351
          Chromosome Location:
          Chr.9: 57129982 - 57131290 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952635_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Man2c1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_102289.1 1 - 2 743 99
          GenBank mRNA AK010793.1 - 1 - 2 701 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI553210.1 - 1 - 2 393 Chr. 9 - 57130023 - 57130926 [-] on Build GRCm38 99
          GenBankmRNA AI481959.1 - 1 - 2 Chr. 9 - 57129983 - 57130730 [+] on Build GRCm38 99
          GenBankmRNA AI851351.1 - 1 - 2 Chr. 9 - 57129987 - 57130747 [+] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transcriptional regulator,
          SIN3A (yeast)
          Sin3a AW553200,
          Sin3,
          mKIAA4126,
          mSin3A
          -1.6kb Anti-sense Chr. 9 - 57071991 - 57128368 on NCBI Build GRCm38
          Downstream nei endonuclease VIII-like 1 (E. coli) Neil1 2810450N13Rik,
          Nei1
          11.5kb Sense Chr. 9 - 57142788 - 57148675 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002364.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2303478 AI851351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2303478 AK010793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2303473 AK077427.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293450 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2303478 AI851351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2303478 AK010793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2303473 AK077427.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01941245 - -
          Whole Mouse Genome G4122A A_52_P244367 AI851351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P244367 AK010793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292665 AK077427.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900019786.mm.1 TC0900002364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019786.mm.1 NR_102289.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019787.mm.1 TC0900002364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019787.mm.1 NR_102289.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019788.mm.1 TC0900002364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019788.mm.1 NR_102289.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019789.mm.1 TC0900002364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019789.mm.1 NR_102289.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 537423 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101400204 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0