Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953004_s1
          See other 2610201A13RIK GE Assays ›
          Gene Symbol:
          2610201A13Rik
          Gene Name:
          RIKEN cDNA 2610201A13 gene
          Gene Aliases:
          AW488471
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953004_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610201A13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AA222883.1 - 1 265 62
          AA473299.1 - 1 265 62
          AK011855.1 - 1 850 62
          AV298639.1 - 1 184 62
          AW488471.1 - 1 62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB085279.1 - 1 225 Chr. 11 - 33150505 - 33150828 [-] on Build GRCm38 62
          GenBankmRNA CA877000.1 - 1 Chr. 11 - 33150508 - 33151110 [+] on Build GRCm38 62
          GenBankmRNA CJ308144.1 - 1 318 Chr. 11 - 33150505 - 33150922 [-] on Build GRCm38 62
          GenBankmRNA BY660605.1 - 1 274 Chr. 11 - 33150505 - 33150878 [-] on Build GRCm38 62
          GenBankmRNA BY660218.1 - 1 264 Chr. 11 - 33150505 - 33150865 [-] on Build GRCm38 62
          GenBankmRNA AV117193.1 - 1 250 Chr. 11 - 33150508 - 33150854 [-] on Build GRCm38 62
          GenBankmRNA BE945772.1 - 1 Chr. 11 - 33150499 - 33150973 [+] on Build GRCm38 62
          GenBankmRNA CJ303826.1 - 1 314 Chr. 11 - 33150505 - 33150918 [-] on Build GRCm38 62
          GenBankmRNA BY668019.1 - 1 290 Chr. 11 - 33150505 - 33150894 [-] on Build GRCm38 62
          GenBankmRNA BX511569.1 - 1 266 Chr. 11 - 33150499 - 33150870 [-] on Build GRCm38 62
          GenBankmRNA BY360958.1 - 1 316 Chr. 11 - 33150501 - 33150918 [-] on Build GRCm38 62
          GenBankmRNA BY661354.1 - 1 269 Chr. 11 - 33150505 - 33150873 [-] on Build GRCm38 62
          GenBankmRNA BM936630.1 - 1 613 Chr. 11 - 33150573 - 33151207 [-] on Build GRCm38 62
          GenBankmRNA BY662930.1 - 1 259 Chr. 11 - 33150505 - 33150863 [-] on Build GRCm38 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream fibroblast growth factor 18 Fgf18 D130055P09Rik,
          FGF-18
          -3.1kb Sense Chr. 11 - 33116970 - 33147400 on NCBI Build GRCm38
          Downstream nucleophosmin 1 Npm1 B23,
          NO38,
          Npm
          1.0kb Sense Chr. 11 - 33152498 - 33163206 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361547 AW488471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361547 AA222883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361547 AA473299.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361547 AK011855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2361547 AV298639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P225370 AW488471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P225370 AK011855.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1420267_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420268_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1420269_PM_at - -
          Mouse Genome 430 2.0 Array 1420268_x_at - -
          Mouse Genome 430 2.0 Array 1420269_at - -
          Mouse Genome 430 2.0 Array 1420267_at - -
          Mouse Genome 430 2.0 Array 1420269_PM_at - -
          Mouse Genome 430 2.0 Array 1420267_PM_at - -
          Mouse Genome 430 2.0 Array 1420268_PM_x_at - -
          Mouse Expression Set 430 1420269_at - -
          Mouse Expression Set 430 1420268_x_at - -
          Mouse Expression Set 430 1420267_at - -
          Mouse Expression Set 430 1420267_PM_at - -
          Mouse Expression Set 430 1420269_PM_at - -
          Mouse Expression Set 430 1420268_PM_x_at - -
          Murine Genome U74v2 Set 95927_PM_f_at - -
          Murine Genome U74v2 Set 161134_PM_at - -
          Murine Genome U74v2 Set 95927_f_at - -
          Murine Genome U74v2 Set 161134_at - -
          Murine Genome U74v2 Set 168120_r_at - -
          Murine Genome U74v2 Set 168120_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1420268_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1420269_at - -
          Mouse Genome 430A 2.0 Array 1420267_at - -
          Mouse Genome 430A 2.0 Array 1420267_PM_at - -
          Mouse Genome 430A 2.0 Array 1420268_x_at - -
          Mouse Genome 430A 2.0 Array 1420269_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 351910 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105890592 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0