Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953079_s1
          See other 2610306M01RIK GE Assays ›
          Gene Symbol:
          2610306M01Rik
          Gene Name:
          RIKEN cDNA 2610306M01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 86848407 - 86849440 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953079_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610306M01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028298.1 1 50 65
          GenBank mRNA AK011997.1 - 1 51 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF583043.1 - 1 51 Chr. 6 - 86848643 - 86849431 [-] on Build GRCm38 65
          GenBankmRNA BY711193.1 - 1 47 Chr. 6 - 86848544 - 86849437 [-] on Build GRCm38 65
          GenBankmRNA CJ182237.1 - 1 47 Chr. 6 - 86848991 - 86849436 [-] on Build GRCm38 65
          GenBankmRNA BB647924.1 - 1 35 Chr. 6 - 86848735 - 86849424 [-] on Build GRCm38 65
          GenBankmRNA CX733586.1 - 1 31 Chr. 6 - 86848798 - 86849421 [-] on Build GRCm38 65
          GenBankmRNA AW495534.1 - 1 258 Chr. 6 - 86849265 - 86849648 [-] on Build GRCm38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream annexin A4 Anxa4 AI265406,
          AIV,
          AW106930,
          Anx4,
          Xanx-4
          -54.8kb Sense Chr. 6 - 86736840 - 86793593 on NCBI Build GRCm38
          Downstream AP2 associated kinase 1 Aak1 5530400K14Rik,
          9630042K20,
          AU067724,
          AU067726,
          BC028270,
          C79663,
          D6Ertd245e,
          R75501,
          mKIAA1048
          42.0 Anti-sense Chr. 6 - 86849482 - 87003227 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002646.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 BC058130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 BC096561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 NR_028298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 AK011997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 AK015614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 BC058130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 BC096561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 NR_028298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 AK011997.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P210340 AK015614.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458212 - -
          Whole Mouse Genome G4122A A_51_P210340 BC058130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 BC096561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 NR_028298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 AK011997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 AK015614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 BC058130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 BC096561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 NR_028298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 AK011997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210340 AK015614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130339 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1452988_PM_at - -
          Mouse Genome 430 2.0 Array 1452988_at - -
          Mouse Genome 430 2.0 Array 1452988_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67170_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67170_at - -
          Affymetrix Mouse Clariom S Array PSR0600020584.mm.1 TC0600002646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020584.mm.1 NR_028298.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020584.mm.1 NR_028298.1 Refseq
          Mouse Expression Set 430 1452988_at - -
          Mouse Expression Set 430 1452988_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452988_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452988_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline