Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953207_s1
          See other 2700008G24RIK GE Assays ›
          Gene Symbol:
          2700008G24Rik
          Gene Name:
          RIKEN cDNA 2700008G24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953207_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2700008G24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK012221.1 - 1 747 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CF894220.1 - 1 252 Chr. 14 - 63664943 - 63665487 [+] on Build GRCm38 90
          GenBankmRNA BY711398.1 - 1 747 Chr. 14 - 63664448 - 63665437 [+] on Build GRCm38 90
          GenBankmRNA BF470400.1 - 1 82 Chr. 14 - 63665113 - 63665671 [+] on Build GRCm38 90
          GenBankmRNA CF892298.1 - 1 252 Chr. 14 - 63664943 - 63665517 [+] on Build GRCm38 90
          GenBankmRNA CF892283.1 - 1 252 Chr. 14 - 63664943 - 63665562 [+] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 15918 Gm15918 OTTMUSG00000027836 -59.7kb Anti-sense Chr. 14 - 63601070 - 63604706 on NCBI Build GRCm38
          Downstream microRNA 598 Mir598 Mirn598,
          mir-598,
          mmu-mir-598
          61.0kb Sense Chr. 14 - 63727189 - 63727267 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2258375 AI448802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2258375 AK012221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2258375 AW495672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2258375 BC038474.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11621035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11621041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11792546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11792556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11707542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29662655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453328_PM_at - -
          Mouse Genome 430 2.0 Array 1453328_PM_at - -
          Mouse Genome 430 2.0 Array 1453328_at - -
          Mouse Expression Set 430 1453328_at - -
          Mouse Expression Set 430 1453328_PM_at - -
          Murine Genome U74v2 Set 133465_at - -
          Murine Genome U74v2 Set 133465_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453328_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453328_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 531883 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100510494 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0