Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953240_s1
          See other CAHM GE Assays ›
          Gene Symbol:
          Cahm
          Gene Name:
          colon adenocarcinoma hypermethylated RNA
          Gene Aliases:
          B930003M22Rik
          Chromosome Location:
          Chr.17: 10319919 - 10321141 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953240_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cahm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037588.1 1 50 94
          GenBank mRNA AK012373.1 - 1 50 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream quaking Qk 1110003F05Rik,
          1500005P18,
          QkI,
          l(17)-1Wis,
          l17Wis1
          -558.0 Anti-sense Chr. 17 - 10202856 - 10319361 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700110C19 gene 1700110C19Rik 3.5kb Sense Chr. 17 - 10324602 - 10329312 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000101.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133549 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101017 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101017 - -
          Whole Mouse Genome G4122A A_52_P472098 AK012373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472098 AK046926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472098 NR_037588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P472098 U44941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154788 AK046926.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101017 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1429748_PM_at - -
          Mouse Genome 430 2.0 Array 1429748_at - -
          Affymetrix Mouse Clariom S Array PSR1700000853.mm.1 TC1700000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000853.mm.1 NR_037588.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000853.mm.1 NR_037588.1 Refseq
          Mouse Expression Set 430 1429748_PM_at - -
          Mouse Expression Set 430 1429748_at - -
          Mouse Genome 430A 2.0 Array 1429748_PM_at - -
          Mouse Genome 430A 2.0 Array 1429748_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 775145 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0