Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953295_s1
          See other DUSP4 GE Assays ›
          Gene Symbol:
          Dusp4
          Gene Name:
          dual specificity phosphatase 4
          Gene Aliases:
          2700078F24Rik, AI844617, BB104621, E130306H24Rik, MKP2
          Chromosome Location:
          Chr.8: 34807610 - 34819894 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953295_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dusp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK012530.1 - 1050 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA EH099308.1 - 1 231 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA BQ554989.1 - 1 Chr. 8 - 34821474 - 34821951 [-] on Build GRCm38 97
          GenBankmRNA EH101990.1 - 1 231 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA BQ746458.1 - 1 383 Chr. 8 - 34821251 - 34821952 [+] on Build GRCm38 97
          GenBankmRNA EH095547.1 - 1 215 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA EH096556.1 - 1 215 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA BP755408.1 - 1 Chr. 8 - 34820818 - 34821951 [-] on Build GRCm38 97
          GenBankmRNA EH095230.1 - 1 215 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA CK622907.1 - 1 221 Chr. 8 - 34821413 - 34821874 [+] on Build GRCm38 97
          GenBankmRNA AA269814.1 - 1 202 Chr. 8 - 34821432 - 34821708 [+] on Build GRCm38 97
          GenBankmRNA EH105555.1 - 1 Chr. 8 - 34821417 - 34821867 [-] on Build GRCm38 97
          GenBankmRNA CK621027.1 - 1 71 Chr. 8 - 34821563 - 34821954 [+] on Build GRCm38 97
          GenBankmRNA BM114666.1 - 1 Chr. 8 - 34821433 - 34821951 [-] on Build GRCm38 97
          GenBankmRNA BF730202.1 - 1 Chr. 8 - 34821384 - 34821956 [-] on Build GRCm38 97
          GenBankmRNA BF714574.1 - 1 278 Chr. 8 - 34821356 - 34821855 [+] on Build GRCm38 97
          GenBankmRNA EH106478.1 - 1 231 Chr. 8 - 34821419 - 34821867 [+] on Build GRCm38 97
          GenBankmRNA BP761364.1 - 1 Chr. 8 - 34821583 - 34821952 [-] on Build GRCm38 97
          GenBankmRNA BY470228.1 - 1 210 Chr. 8 - 34821423 - 34821947 [+] on Build GRCm38 97
          GenBankmRNA AA033135.1 - 1 218 Chr. 8 - 34821416 - 34821944 [+] on Build GRCm38 97
          GenBankmRNA BF321109.1 - 1 Chr. 8 - 34821498 - 34821952 [-] on Build GRCm38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Downstream tankyrase,
          TRF1-interacting ankyrin-related ADP-ribose polymerase
          Tnks 4930554K12Rik,
          AI662855,
          ARTD5,
          C86528,
          D130072O21Rik,
          TANK1,
          mTNKS1
          7.3kb Anti-sense Chr. 8 - 34829179 - 34965690 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix