Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954085_s1
          See other 2900054C01RIK GE Assays ›
          Gene Symbol:
          2900054C01Rik
          Gene Name:
          RIKEN cDNA 2900054C01 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954085_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2900054C01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK013682.1 - 1 1053 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB009928.1 - 1 218 Chr. X - 160801627 - 160801899 [-] on Build GRCm38 85
          GenBankmRNA BB176149.1 - 1 208 Chr. X - 160801627 - 160801907 [-] on Build GRCm38 85
          GenBankmRNA BM202541.2 - 1 Chr. X - 160801625 - 160802129 [+] on Build GRCm38 85
          GenBankmRNA BQ176056.1 - 1 Chr. X - 160801392 - 160802078 [+] on Build GRCm38 85
          GenBankmRNA BQ557839.1 - 1 Chr. X - 160801625 - 160802249 [+] on Build GRCm38 85
          GenBankmRNA BY592462.1 - 1 Chr. X - 160801394 - 160801857 [-] on Build GRCm38 86
          GenBankmRNA CJ261452.1 - 1 391 Chr. X - 160801629 - 160802090 [-] on Build GRCm38 85
          GenBankmRNA AW324348.1 - 1 Chr. X - 160801626 - 160801963 [+] on Build GRCm38 85
          GenBankmRNA BB240852.1 - 1 225 Chr. X - 160801628 - 160801924 [-] on Build GRCm38 85
          GenBankmRNA BM935238.1 - 1 Chr. X - 160801600 - 160801811 [+] on Build GRCm38 85
          GenBankmRNA BB429508.1 - 1 218 Chr. X - 160801625 - 160801917 [-] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream retinoschisis (X-linked,
          juvenile) 1 (human)
          Rs1 Rs1h,
          Xlrs1,
          tmgc1
          -1.7kb Anti-sense Chr. X - 160768013 - 160799663 on NCBI Build GRCm38
          Downstream gap junction protein,
          alpha 6
          Gja6 A530015O12Rik,
          Cnx33,
          Cx33,
          Gja-6
          99.4kb Sense Chr. X - 160902116 - 160907052 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2217831 AK013682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2217831 AK013682.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429625_PM_at - -
          Mouse Genome 430 2.0 Array 1429625_PM_at - -
          Mouse Genome 430 2.0 Array 1429625_at - -
          Mouse Expression Set 430 1429625_PM_at - -
          Mouse Expression Set 430 1429625_at - -
          Affymetrix HT Mouse Genome 430B Array 1429625_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429625_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102760121 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0