Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954292_s1
          See other 2900076G11RIK GE Assays ›
          Gene Symbol:
          2900076G11Rik
          Gene Name:
          RIKEN cDNA 2900076G11 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954292_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2900076G11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK013797.1 - 1 347 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream expressed sequence AI839979 AI839979 -9.7kb Anti-sense Chr. 5 - 31569592 - 31571397 on NCBI Build GRCm38
          Downstream predicted gene,
          38424
          Gm38424 14.0kb Sense Chr. 5 - 31595752 - 31663432 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04682800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04728354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04219964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04728357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04573295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04889136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04715290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04728358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04728360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04573299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04682802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04784364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23578872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04784366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04889135 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 168178_PM_at - -
          Murine Genome U74v2 Set 168178_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix