Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954520_sH
          See other 3110009M11RIK GE Assays ›
          Gene Symbol:
          3110009M11Rik
          Gene Name:
          RIKEN cDNA 3110009M11 gene
          Gene Aliases:
          D7Ssb2
          Chromosome Location:
          Chr.7: 83864498 - 83866392 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954520_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 3110009M11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK014034.1 - 1 51 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY713034.1 - 1 51 Chr. 7 - 83864340 - 83865291 [-] on Build GRCm38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700026D08 gene 1700026D08Rik AW047638,
          AW551802
          -69.0kb Sense Chr. 7 - 83774101 - 83795384 on NCBI Build GRCm38
          Downstream mesoderm development candidate 1 Mesdc1 AW061151 15.2kb Sense Chr. 7 - 83880495 - 83884341 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P06251823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06487120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06266188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06266185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06441301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06646374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06251826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06170613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25285111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06527260 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 592173 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105130059 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0