Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954661_s1
          See other 4632427E13RIK GE Assays ›
          Gene Symbol:
          4632427E13Rik
          Gene Name:
          RIKEN cDNA 4632427E13 gene
          Gene Aliases:
          AU015482
          Chromosome Location:
          Chr.7: 92740706 - 92741459 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954661_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4632427E13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK014157.1 - 1546 91
          AK019505.1 - 1577 91
          AK051992.1 - 1565 91
          AK084084.1 - 1582 91
          AK145781.1 - 1587 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK343135.1 - 1 77 Chr. 7 - 92739352 - 92739959 [-] on Build GRCm38 91
          GenBankmRNA AU015482.1 - 1 Chr. 10 - 29345520 - 29345951 [-] on Build GRCm38 91
          GenBankmRNA BG070284.2 - 1 Chr. 7 - 92739624 - 92740078 [+] on Build GRCm38 91
          GenBankmRNA CJ283271.1 - 1 126 Chr. 7 - 92739626 - 92740008 [-] on Build GRCm38 91
          GenBankmRNA CK342659.1 - 1 77 Chr. 7 - 92739310 - 92739959 [-] on Build GRCm38 91
          GenBankmRNA BY682521.1 - 1 140 Chr. 7 - 92739626 - 92740021 [-] on Build GRCm38 91
          GenBankmRNA CK342806.1 - 1 77 Chr. 7 - 92739290 - 92739959 [-] on Build GRCm38 91
          GenBankmRNA CK343195.1 - 1 77 Chr. 7 - 92739281 - 92739959 [-] on Build GRCm38 91
          GenBankmRNA CJ283258.1 - 1 122 Chr. 7 - 92739626 - 92740004 [-] on Build GRCm38 91
          GenBankmRNA BB681022.1 - 1 227 Chr. 7 - 92739629 - 92740109 [-] on Build GRCm38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cleavage and polyadenylation factor subunit homolog (S. cerevisiae) Pcf11 2500001H09Rik,
          5730417B17Rik,
          C77803
          -69.5kb Anti-sense Chr. 7 - 92643458 - 92670087 on NCBI Build GRCm38
          Downstream R-spondin 3 homolog (Xenopus laevis) Rspo3 2810459H04Rik,
          AW742308,
          Cristin1,
          Thsd2
          107.2kb Sense Chr. 10 - 29453106 - 29535867 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0