Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921520E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014928.1 | - | 1 | 550 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ets variant 2 | Etv2 | Etsrp71 | -121.0 | Anti-sense | Chr. 7 - 30633616 - 30636509 on NCBI Build GRCm38 |
Downstream | RNA binding motif protein 42 | Rbm42 |
1700003D06Rik, 3100004P22Rik |
2.6kb | Anti-sense | Chr. 7 - 30640995 - 30650228 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |