Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03955741_s1
          See other 4930402C16RIK GE Assays ›
          Gene Symbol:
          4930402C16Rik
          Gene Name:
          RIKEN cDNA 4930402C16 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03955741_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930402C16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015044.1 - 1 1248 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 14025 Gm14025 OTTMUSG00000015417 Anti-sense Chr. 2 - 129024141 - 129065905 on NCBI Build GRCm38
          Downstream tubulin tyrosine ligase Ttl 2410003M22Rik,
          2700049H19Rik,
          AI848570
          26.0kb Sense Chr. 2 - 129065947 - 129096283 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01299557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01299555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01387286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01587989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01917918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01970344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01587983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01587977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01587963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21622687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1433084_PM_at - -
          Mouse Genome 430 2.0 Array 1433084_at - -
          Mouse Genome 430 2.0 Array 1454467_PM_at - -
          Mouse Genome 430 2.0 Array 1454467_at - -
          Mouse Genome 430 2.0 Array 1433084_PM_at - -
          Mouse Expression Set 430 1454467_PM_at - -
          Mouse Expression Set 430 1433084_PM_at - -
          Mouse Expression Set 430 1454467_at - -
          Mouse Expression Set 430 1433084_at - -
          Affymetrix HT Mouse Genome 430B Array 1433084_at - -
          Affymetrix HT Mouse Genome 430B Array 1433084_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0