Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03955977_s1
          See other 4930425P05RIK GE Assays ›
          Gene Symbol:
          4930425P05Rik
          Gene Name:
          RIKEN cDNA 4930425P05 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03955977_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930425P05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015204.1 - 1 653 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714759.1 - 1 653 Chr. 14 - 32012173 - 32012942 [-] on Build GRCm38 102
          GenBankmRNA CJ189643.1 - 1 261 Chr. 14 - 32012141 - 32012545 [-] on Build GRCm38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ankyrin repeat domain 28 Ankrd28 AI465466,
          AI604979,
          E430019N21Rik,
          mKIAA0379
          -181.7kb Sense Chr. 14 - 31698768 - 31830415 on NCBI Build GRCm38
          Downstream UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15 Galnt15 4631401E18Rik,
          Galntl2,
          mpp-GalNAc-T15
          15.7kb Anti-sense Chr. 14 - 32028651 - 32062204 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P11329794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11596816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11596814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11596809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11404614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11315987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11596818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11481639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11596811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11315983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11807257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11315982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11292147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11264904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11148536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11180304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515075 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0