Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956055_g1
          See other 4930429P21RIK GE Assays ›
          Gene Symbol:
          4930429P21Rik
          Gene Name:
          RIKEN cDNA 4930429P21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 107348949 - 107354183 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956055_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930429P21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015246.1 - 1 - 2 249 129
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714803.1 - 1 - 2 249 Chr. 9 - 107353289 - 107353986 [-] on Build GRCm38 129

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 6430571L13 gene 6430571L13Rik -3.6kb Anti-sense Chr. 9 - 107340640 - 107349683 on NCBI Build GRCm38
          Downstream calcium channel,
          voltage-dependent,
          alpha 2/delta subunit 2
          Cacna2d2 Cacna2d,
          a2d2,
          du,
          mKIAA0558,
          td,
          torpid
          45.9kb Anti-sense Chr. 9 - 107399880 - 107529346 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788733 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08136911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07678918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26808043 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 391979 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0