Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956276_s1
          See other 0610009F21RIK GE Assays ›
          Gene Symbol:
          0610009F21Rik
          Gene Name:
          RIKEN cDNA 0610009F21 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956276_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 0610009F21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK002391.1 - 1 352 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY702926.1 - 1 350 Chr. 16 - 91947327 - 91947788 [+] on Build GRCm38 64
          GenBankmRNA AV000737.1 - 1 279 Chr. 16 - 91947396 - 91947786 [+] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP synthase,
          H+ transporting,
          mitochondrial F1 complex,
          O subunit
          Atp5o ATPO,
          D12Wsu28e,
          OSCP
          -15.7kb Anti-sense Chr. 16 - 91925223 - 91931630 on NCBI Build GRCm38
          Downstream solute carrier family 5 (inositol transporters),
          member 3
          Slc5a3 AA623876,
          BF642829,
          Smit1
          110.5kb Sense Chr. 16 - 92058322 - 92087473 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12750004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12850578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12902198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30966123 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104670017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0