Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956286_m1
          See other 0610031O16RIK GE Assays ›
          Gene Symbol:
          0610031O16Rik
          Gene Name:
          RIKEN cDNA 0610031O16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 138210717 - 138239663 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956286_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 0610031O16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045760.1 2 - 3 286 86
          GenBank mRNA AK002733.1 - 2 - 3 286 86

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream alcohol dehydrogenase 7 (class IV),
          mu or sigma polypeptide
          Adh7 AI325182,
          Adh-3,
          Adh-3e,
          Adh-3t,
          Adh3,
          Adh3-e,
          Adh3-t,
          Adh4,
          Adt-1
          Anti-sense Chr. 3 - 138198053 - 138232042 on NCBI Build GRCm38
          Downstream alcohol dehydrogenase 1 (class I) Adh1 ADH-AA,
          AI194826,
          Adh-1,
          Adh-1-t,
          Adh-1e,
          Adh-1t,
          Adh-3e,
          Adh1-e,
          Adh1-t,
          Adh1tl,
          Adh3-e
          39.9kb Anti-sense Chr. 3 - 138277645 - 138290691 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003018.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111591 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00768704 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106205 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02602709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02939438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03023523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22567793 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300023819.mm.1 TC0300003018.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300023819.mm.1 NR_045760.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300023819.mm.1 NR_045760.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0