Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956406_s1
          See other 1110007E10RIK GE Assays ›
          Gene Symbol:
          1110007E10Rik
          Gene Name:
          RIKEN cDNA 1110007E10 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956406_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1110007E10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003532.1 - 1 747 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV007945.1 - 1 136 Chr. 4 - 124640533 - 124640837 [+] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 2164 Gm2164 Anti-sense Chr. 4 - 124522925 - 124657504 on NCBI Build GRCm38
          Downstream POU domain,
          class 3,
          transcription factor 1
          Pou3f1 Oct-6,
          Oct6,
          Otf-6,
          Otf6,
          Scip,
          Test1,
          Tst-1,
          Tst1
          16.8kb Sense Chr. 4 - 124657646 - 124660655 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777346 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03973741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03736890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03471413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03237248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03680317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03372303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04087989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03471410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03736892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03320131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03237249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23269372 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103850592 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0