Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1439541_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1456636_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1447114_PM_x_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1443790_PM_x_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1456779_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1439541_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456636_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456779_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1447114_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1439541_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456636_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1447114_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1443790_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1443790_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1456779_PM_a_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0600006517.mm.1 |
TC0600000851.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006517.mm.1 |
NR_046011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006517.mm.1 |
NR_046011.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006516.mm.1 |
TC0600000851.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006516.mm.1 |
NR_046011.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006516.mm.1 |
NR_046011.1 |
Refseq |
Mouse Expression Set 430 |
1456636_at |
- |
- |
Mouse Expression Set 430 |
1456636_PM_at |
- |
- |
Mouse Expression Set 430 |
1443790_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1456779_PM_a_at |
- |
- |
Mouse Expression Set 430 |
1456779_a_at |
- |
- |
Mouse Expression Set 430 |
1447114_x_at |
- |
- |
Mouse Expression Set 430 |
1439541_PM_at |
- |
- |
Mouse Expression Set 430 |
1439541_at |
- |
- |
Mouse Expression Set 430 |
1443790_x_at |
- |
- |
Mouse Expression Set 430 |
1447114_PM_x_at |
- |
- |
Murine Genome U74v2 Set |
171260_PM_at |
- |
- |
Murine Genome U74v2 Set |
167564_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
171260_at |
- |
- |
Murine Genome U74v2 Set |
167564_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456636_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1456636_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456636_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1456636_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1456779_PM_a_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439541_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1443790_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1447114_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1443790_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1447114_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439541_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1456779_a_at |
- |
- |