Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03957268_s1
          See other 4930440F04RIK GE Assays ›
          Gene Symbol:
          4930440F04Rik
          Gene Name:
          RIKEN cDNA 4930440F04 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03957268_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930440F04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019603.1 - 1 751 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930465M20 gene 4930465M20Rik 4930431H15Rik -163.6kb Anti-sense Chr. 12 - 107723743 - 107735488 on NCBI Build GRCm38
          Downstream B cell leukemia/lymphoma 11B Bcl11b 9130430L19Rik,
          AI604821,
          B630002E05Rik,
          BCL-11B,
          Ctip2,
          Rit1
          10.5kb Anti-sense Chr. 12 - 107910403 - 108004139 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09955074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09955067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09955064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09847394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09955063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09847396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10197710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28724329 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103170520 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0