Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03957403_s1
          See other 4930564I24RIK GE Assays ›
          Gene Symbol:
          4930564I24Rik
          Gene Name:
          RIKEN cDNA 4930564I24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03957403_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930564I24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019783.1 - 1 251 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ205012.1 - 377 Chr. - - [-] on Build 65
          GenBankmRNA BY715829.1 - 1 251 Chr. 2 - 35979929 - 35980248 [+] on Build GRCm38 65
          GenBankmRNA BY715837.1 - 1 251 Chr. 2 - 35979929 - 35980242 [+] on Build GRCm38 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tubulin tyrosine ligase-like family,
          member 11
          Ttll11 4932702F08Rik,
          4933424A20Rik,
          AU043211,
          D2Ertd624e
          -305.0 Anti-sense Chr. 2 - 35751226 - 35979624 on NCBI Build GRCm38
          Downstream NADH dehydrogenase (ubiquinone) 1 alpha subcomplex,
          8
          Ndufa8 0610033L03Rik,
          AW261656
          56.1kb Anti-sense Chr. 2 - 36036334 - 36049292 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02021109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02245654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01900560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01292023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02245651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01817813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02245658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01817814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02111258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01817817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21137065 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 649714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0